Dropping funnels, cylindrical, without pressure balance
Supplier: Lenz Laborglas GmbH & CO.KG
Certificates
About this item
DURAN®, borosilicate glass, clear, with glass key.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Standard ground joint cone and socket. With divisions. Stopcock with thread guard.