Petri dishes, Corning® Gosselin™
Supplier: Corning
Certificates
About this item
Transparent PS Petri dishes for food industry laboratories.
- Excellent optical quality
- Stable stacking due to the stacking ring
- High level of mechanical resistance, will retain shape up to +55 °C
These dishes are produced in a Class 100 (ISO 5) manufacturing environment, ensuring an aseptic product. Product is labelled with a batch number on each carton. Also available sterile (Sterility Assurance Level based on ISO 11137 : SAL 10 ⁻⁶).
Specifications
- Catalog No:
- GOSSBP143-07
- 391-0569
- GOSSBP143-06
- GOSSBP93B-102
- GOSSBP93D-102
- 391-0566
- 391-0570
- 391-0567
- 391-0568
- GOSSBP93B-101
- 391-0572
- GOSSSB93-102
- 391-0571
- GOSSBP90B-102
- GOSSBP140-02
- 391-2068
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- RECs, ISO 14001 - Borre
- RECs, ISO 14001 - Borre
- RECs, ISO 14001 - Borre
- RECs, ISO 14001 - Borre
- RECs, ISO 14001 - Borre
- RECs, ISO 14001 - Borre
- RECs, ISO 14001 - Borre
- RECs, ISO 14001 - Borre
- RECs, ISO 14001 - Borre
- RECs, ISO 14001 - Borre
- RECs, ISO 14001 - Borre
- RECs, ISO 14001 - Borre
- RECs, ISO 14001 - Borre
- RECs, ISO 14001 - Borre
- RECs, ISO 14001 - Borre
- RECs, ISO 14001 - Borre