VWR® Hichrom Fittings
Supplier: Avantor
Certificates
About this item
PEEK. Can be used at temperatures up to 150 °C. Unlike stainless steel fittings, PEEK fingertight fittings will not permanently lock into place on the tubing. Compatible with all types of end fittings.
- Fingertight to 6000 psi
- Void-free connection
- Inert and biocompatible