Anion exchange chiral stationary phases
For the separation of acidic chiral compounds containing carboxylic, phosphonic, phosphinic, phosphoric or sulphonic acid groups. In some cases weakly acidic compounds, such as phenols, can also be separated.
For the separation of acidic chiral compounds containing carboxylic, phosphonic, phosphinic, phosphoric or sulphonic acid groups. In some cases weakly acidic compounds, such as phenols, can also be separated.