Trousers, Tyvek® 500, model PT31L0
: DuPont
Specially designed for use with Tyvek® apparel, these trousers can help offer enhanced protection for body parts that are more exposed to hazardous substances. Tyvek® 500 trousers are composed of flash spun high density polyethylene, providing an ideal balance of protection, durability and comfort. Tyvek® is permeable to both air and water vapour, yet repels water-based liquids and aerosols. It offers an excellent barrier against fine particles and fibres (down to 1 µm in size).
- Fabric is ultra-low linting
- Without pockets
- Elasticated waist, no elastic at ankles
- Stitched internal seams
Applications include pharmaceutical handling, chemical protection, lead and asbestos abatement/remediation, general maintenance/operations, spray painting and general clean-up, amongst many others.
Colour: White
Weight: 41 g/m²
: (EU) 2016/425; Cat III, Type PB [6-B]; EN 14126
: 50 per box, bulk packed.
- Catalog No:
- 113-0484
- 113-0482
- 113-0483
- 113-0481
- Model:
- 500 PT31L0
- 500 PT31L0
- 500 PT31L0
- 500 PT31L0
- Garment application:
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Risk area:
- Anti-static
- Anti-static
- Anti-static
- Anti-static
- Gender:
- Unisex
- Unisex
- Unisex
- Unisex
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Fabric:
- Tyvek®
- Tyvek®
- Tyvek®
- Tyvek®
- Weight:
- 41 g/m²
- 41 g/m²
- 41 g/m²
- 41 g/m²