Order Entry
Finland
ContactUsLinkComponent
Tubing clamps, Kocher (Ochsner) type
Tubing clamps, Kocher (Ochsner) type
  231-0251
undefined
Tubing clamps, Kocher (Ochsner) type
  231-0251
  • Length:
    200 mm
  • Pk:
    1
Stainless steel, straight. Ideal for stopping or controlling liquid flow rates in tubing connections.
  • Very good thermal resistance, autoclavable
  • Corrosion-resistant
  • Antimagnetic