Burette clamps, double
Catalog # 241-3169
Supplier: BOCHEM
Specifications
- For:Two burettes
- Description:Clamp for 2 burettes
- Pk:1
Specifications
About this item
Aluminium.
- With bosshead for fixing to retort stand with Ø 0 to 13
- With PVC rolls
- Distance between rolls 170 mm