Burettes, lateral stopcock, Schellbach, BLAUBRAND®
Supplier: Brand
Certificates
About this item
Borosilicate glass 3.3, class AS, DE-M marking.
- Calibrated to deliver 'TD, Ex'
- Schellbach stripe, ring marks at major graduations
- Marks and inscriptions in high contrast blue enamel represents the optimum combination of resistance and readability
Length: 800 mm.
Length: 800 mm
Certifications: DIN EN ISO 385
Ordering information: Supplied with a batch certificate. Burettes are also available on request with an individual certificate, a USP individual certificate or a DAkkS calibration certificate.