Biohazard incinerator cartons, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
Certificates
About this item
Corrugated cardboard.
- Snug fitting lid has flap through which waste is inserted
- When carton is filled, internal safety cap is pulled into place and the entire unit is ready for incineration
Carton with integral biohazard bag in HDPE.
Delivery: Delivered unassembled. Assembly takes a few seconds.