Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Closures
Bottle Closures
Screw caps for wide neck bottles, rectangular
Screw caps for wide neck bottles, rectangular
215-3050
:
MELLERUD PLAST EMBALLATOR
Screw caps for wide neck bottles, rectangular
215-3050
:
MELLERUD PLAST EMBALLATOR
Thread:
100 mm
Colour:
Black
Material:
PE
Pk:
1