Bottles, narrow neck, with standard ground stoppers
Supplier: Kavalier
Certificates
About this item
Soda-lime glass, clear or amber, with standard ground joint and glass stopper.
- Good chemical and physical properties
- For use in applications without high thermal loads
- Solid, replaceable glass stoppers
Amber to protect light-sensitive materials.
DIN 12036, ISO 4796
Certifications: DIN 12036, ISO 4796