Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Closures
Bottle Closures
Caps for narrow neck aluminium bottles
Caps for narrow neck aluminium bottles
:
TOURNAIRE-EMBALLAGE
Caps for narrow neck aluminium bottles
:
TOURNAIRE-EMBALLAGE
PP or PE, white.
Only suitable for 50 – 4000 ml bottles
Aluthene insert ensures absolute leakproof properties, even with aggressive media
Tamper-evident cap for safety and control