Bottle, narrow neck, with thread
Supplier: REXAM
Certificates
About this item
Soda-lime glass, brown, narrow without screw caps.
- Good chemical and physical properties
- For use in applications without high thermal loads
DIN thread. Brown coloured to protect light-sensitive materials and supplied with dust protection cap. Space-saving (square shape) for storage and transport. Stable, non-slip.
Closures in various designs are available on request (see Cat. No. 217-9200 to 217-9203 for screw caps and 217-0202 to 217-0205 for pouring rings)).