Jerricans, narrow neck, HDPE, transparent or blue
Supplier: Kautex
Certificates
About this item
HDPE, natural colour.
- Jerrycans 2,5 to 5 L and 10 to 30 L with UN approval
- Comprehensive range of closures
- Varied use
- Leakproof
- Jerrycans are suitable for food
- Halal certified
Universal jerrycan for industry, trade, and laboratories. These jerrycans, with a comprehensive range of closures accessories, can be used for almost any purpose.