Gas washing bottles, Drechsel pattern, VitraPOR®
Supplier: ROBU GLASFILTER
Certificates
About this item
DURAN®, borosilicate glass, clear, with PBT screw caps.
- Sintered glass filter porosity 1 (sinter/disc diameter 25 mm)
- All system components can be ordered separately
- Excellent chemical resistance and temperature resistance
Comprises of gas washing bottles with screw thread DIN GL 45, versatile use - screw cap and 2 tube connectors made from PP with threaded connection.