Carboys, narrow neck, with screw cap
: KARTELL
HDPE, translucent with blue screw cap.
- Good chemical resistance
- Heat resistant up to 80 °C (120 °C for short periods)
- Heavy walled, very tough, a deeper closure with insert seal increasing leakproof performance
Ideal for storing and transporting liquids. Includes a carrying handle, 25 and 50 litre sizes have two handles.