Jerricans, narrow neck, HDPE
Supplier: MELLERUD PLAST EMBALLATOR
Certificates
About this item
HDPE Bottle with tight-fit lid, moulded handle and lid with locking ring.
Models ranging from 2.5 to 25 litres. Models ranging from 5 to 25 litres are approved for the transportation of hazardous materials. Supplied with screw caps.