Bottle/carboys, rectangular, with handles, HDPE
HDPE, with or without screw cap. Rectangular, with handles. Threaded, 50 and 100 mm.
: The 5 and 10 liter jugs with 100mm opening are intended for use with dry products. If the jug is to be used with liquid, a foil or similar material must be used to open the bottle. Applies to product numbers MELP48C0500100117D and 215-3064.