Order Entry
Finland
ContactUsLinkComponent
Film, Kodak BioMax® MR
Film, Kodak BioMax® MR
Catalog # 730-3245
Supplier:  CARESTREAM HEALTH
undefined
Film, Kodak BioMax® MR
Catalog # 730-3245
Supplier:  CARESTREAM HEALTH
Specifications
  • Dimensions:
    330×610 mm
  • Seq:
    07
  • Pk:
    50
Specifications
About this item
BioMax® MR is a single emulsion film that provides maximum resolution and sensitivity for detection of ³⁵S, ³²P,and ¹⁴C labelled samples. An emulsion coated on a single side of a clear support produces maximum resolution and background clarity in the detection of all labels, making overall reading of sequencing gels easier and more accurate. BioMax® MR film also provides maximum resolution for direct detection of ³²P labelled samples, delivering sharper, crisper band responses that allow viewing in exceptionally fine detail. BioMax® MR film is also suitable for detection of most chemiluminescent labelled samples. Though not as sensitive as BioMax® MS, BioMax® MR film offers maximum resolution and is the recommended film for Western chemiluminescent application where exposures are relatively short.
  • Maximum resolution and background clarity for easy and accurate reading of sequencing gels
  • Recommended for ³⁵S, ³²P and ¹⁴C labelled samples
  • T-Grain emulsion technology enhances sensitivity by two to four times that of other films, giving greater detection for low intensity bands
  • Suitable for manual or automated processing
Packaging: Non-interleaved sheets, 50 per box.