Aluminium foils rolls and dispenser
Supplier: Korff
Certificates
About this item
Pure aluminium, 99,0 to 99,5%, soft.
- Different widths and thicknesses available
- Comes in a practical dispenser
- Economical large size rolls for use in a roll dispenser
Ideal for packaging, covering and insulation.