Order Entry
Finland
ContactUsLinkComponent
Pararosaniline hydrochloride, Certistain® mikroskopiaan, Sigma-Aldrich®
Health
Pararosaniline hydrochloride, Certistain® mikroskopiaan, Sigma-Aldrich®
Catalog # 1.07509.5000
Supplier:  Merck
CAS Number:  
Health
Pararosaniline hydrochloride, Certistain® mikroskopiaan, Sigma-Aldrich®
Catalog # 1.07509.5000
Supplier:  Merck
CAS Number:  

Specifications

  • Pakkaustyyppi:
    Cardboard carton
  • Pak.:
    5 kg
  • Seq:
    4114

Specifications

About this item

 :
 :323.82 g/mol
Melting Pt:268 °C
EINECS:209-321-2
 :GHS08
Storage Temperature:Huoneenlämpö
 :1
CAS Number:569-61-9
Formula:C₁₉H₁₈ClN₃
MDL Number:MFCD00001657