Thymolphthalein
Glass bottle for solids, 100 g
Catalog # MOLE40922033-100G
Marketsource
Supplier: Molekula
Jump to:
Product Details & Documents
Synonyms: 3,3-Bis(4-hydroxy-5-isopropyl-o-tolyl)phthalide, 3,3-Bis(4-hydroxy-5-isopropyl-2-methylphenyl)isobenzofuran-1(3H)-one
- CAS Number:125-20-2
- Formula:C₂₈H₃₀O₄
- MDL Number:MFCD00005909
- Molecular Weight:430.54 g/mol
- Merck Index:12,09542
- Melting Pt:253 °C
- Storage Temperature:Ambient
- EINECS:204-729-7
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g



