Xylenol orange tetrasodium salt indicator for metal titration, Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:3618-43-7
:313223-06-2
Formula:C₃₁H₂₈N₂Na₄O₁₃S
MDL Number:MFCD00044293
:760.59 g/mol
Melting Pt:195 °C
Storage Temperature:Ambient
EINECS:222-805-8
Specification Test Results
- Identity (IR):complying
- Change range:complying
- Sulfated ash:Max. 50 %
- Absorption maximum:575 - 585 nm
- Absorbtion (1%/1cm):Min. 400
- Suitability for Complexometry:complying
- Test of sensitivity:complying