Order Entry
Finland
ContactUsLinkComponent
Light green SF yellowish, Sigma-Aldrich®
Light green SF yellowish, Sigma-Aldrich®
Catalog # L1886-25G
CAS Number:  
undefined
Light green SF yellowish, Sigma-Aldrich®
Catalog # L1886-25G
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :792.86 g/mol
Merck Index:13,05506
Melting Pt:288…289 °C
 :M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:225-906-5
CAS Number:5141-20-8
Formula:C₃₇H₃₄N₂Na₂O₉S₃
MDL Number:MFCD00012121