Methyl blue, pure, certified
Catalog # ACRO401180250
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:Dark brown Glistening crystalline powder or crystals
- Infrared spectrum:Conforms
- UV Lambda max:595 to 605 nm (in 0.005M HCl)
- Absorption Ratio P-15/P+15:0.96 to 1.05
- Titration with TiCl3:≥12.0 ml 0.1N TiCl3/g
- BSC Certification:Passes
Specification Test Results
About this item
:
:799.81 g/mol
EINECS:249-113-9
:GHS07
Storage Temperature:Ambient
CAS Number:28983-56-4
Formula:C₃₇H₂₇N₃Na₂O₉S₃
MDL Number:MFCD00058509