Methyl blue, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Certificates
About this item
CAS Number:28983-56-4
:
Formula:C₃₇H₂₇N₃Na₂O₉S₃
MDL Number:MFCD00058509
Merck Index:13,06055
:M-Clarity™ quality level = MQ200
Flash Pt:>300 °C (Pensky-Martens) (closed cup)
:799.81 g/mol
Storage Temperature:Ambient
EINECS:249-352-9
Specification Test Results
- Appearance (colour):Purple to very dark purple
- Appearance (form):Powder or crystals
- Thin layer chromatogr.:Corresponds to standard
- Solubility (colour):0.01 g in 10 ml water
- Solubility (method):0.01 g in 10 ml water
- pH-transition:9.4
- Colour:Blue
- Wavelength (1) (UV):595.0 - 605.0 nm
- Extinction (1%/1 cm) (1) (UV):≥400
- Wavelength (2) (UV):310.0 - 320.0 nm
- Extinction (1%/1 cm) (2) (UV):≥200
- Solvent (UV):HCl 0.005 m