Human/Mouse/Rat Recombinant GDF-11 (Animal free) (from E. coli)
Supplier: Shenandoah Biotechnology
Certificates
About this item
Growth differentiation factor 11 (GDF-11), also known as bone morphogenetic protein 11 (BMP-11), is a regulator of cell growth and differentiation during muscular and neural development. GDF-11 binds the transforming growth factor-beta receptors ALK4, ALK5, and ALK7 to activate SMAD signaling. In adults, exogenous GDF-11 promotes cardiomyocyte regeneration to reverse age-related cardiac hypertrophy.
- High Quality
- Low Endotoxin
- Affordable
Caution: For research use only.
Specifications
- Conjugation:Unconjugated
- Protein function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:HumanRatMouse
- Storage conditions:At or below –20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤0,1
- Biological activity:Bioactivity assay: Alkaline phosphatase activity in ATDC5 cells, ED50 ≤100 ng/ml, Bioactivity ≥1,0×10⁴ units/mg
- Gene ID:O95390 / Q9Z1W4 / Q9Z217
- Reconstitution instructions:Sterile 10 mM HCl at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:Y
- Protein synonyms:BMP-11
- Protein/peptide name:GDF-11
- Purity:≥95%
- Molecular weight:12,5/24,9 kDa
- Sequence:NLGLDCDEHS SESRCCRYPL TVDFEAFGWD WIIAPKRYKA NYCSGQCEYM FMQKYPHTHL VQQANPRGSA GPCCTPTKMS PINMLYFNDK QQIIYGKIPG MVVDRCGCS
- Endotoxin level:Low
- Formulation:0,1% Trifluoroacetic Acid (TFA)
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
- Tested applications:Bioassay