Order Entry
Finland
ContactUsLinkComponent
 
Human Recombinant IL-2 (CTGrade GMP) (from E. coli)
New Product
 
Human Recombinant IL-2 (CTGrade GMP) (from E. coli)
New Product

 

  • Conjugation:
    Unconjugated
  • Protein function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    At or below –20 °C
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤0,01
  • Biological activity:
    Bioactivity assay: Proliferation of CTLL- 2, ED50 ≤3 ng/ml, Bioactivity ≥1,8×10⁴ IU/µg
  • Gene ID:
    P60568
  • Amino acid number:
    134 aa
  • Reconstitution instructions:
    Sterile 10 mM HCl at 0.1 - 0.5 mg/ml
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    Y
  • Protein synonyms:
    Aldesleukin|TCGF|T cell growth factor
  • Protein/peptide name:
    IL-2 C126S
  • Purity:
    ≥97%
  • Molecular weight:
    15,5 kDa
  • Sequence:
    MAPTSSSTKK TQLQLEHLLL DLQMILNGIN NYKNPKLTRM LTFKFYMPKK ATELKHLQCL EEELKPLEEV LNLAQSKNFH LRPRDLISNI NVIVLELKGS ETTFMCEYAD ETATIVEFLN RWITFSQSII STLT
  • Endotoxin level:
    Low
  • Formulation:
    0,1% Trifluoroacetic Acid (TFA)
  • Nuclease-free:
    N
  • Grade:
    CTGrade GMP
  • Shipping temperature:
    Ambient
  • Tested applications:
    Bioassay