Order Entry
Finland
Orders LinkContactUsLinkComponent
TOCTAX Calibrating Solution

TOCTAX Calibrating Solution

TOCTAX Calibrating solution, 200 mg/L, 1 L
Catalog # HACHLCW845
Supplier:   Hach
TOCTAX Calibrating Solution
Catalog # HACHLCW845
Supplier:  Hach
CAS Number:  
TOCTAX Calibrating Solution

TOCTAX Calibrating solution, 200 mg/L, 1 L

CAS Number:  
Price
143,00 €
Log in for availability
1 * 1 L
Product Details & Documents
Specifications

Specifications

  • Concentration:
    200 mg/L
  • Pk:
    1 L
  • Description:
    TOCTAX Calibrating solution

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.