Starch 1% in aqueous solution stabilised Reag. Ph. Eur. 1085103
Catalog # 85964.180
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:100 mL
- Seq:2440
Specifications
About this item
:
:Stabilised with mercury (II) iodide 0.01 %
:162.14 g/mol
Storage Temperature:Ambient
EINECS:232-679-6
CAS Number:9005-25-8
Formula:(C₆H₁₀O₅)n
MDL Number:MFCD00082026