Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Silica gel
Silica gel, Pore size: 60 Å, Supelco®
Silica gel, Pore size: 60 Å, Supelco®
Silica gel, Pore size: 60 Å, Supelco®
Supplier:
Merck
Certificates
Expand image
Silica gel, Pore size: 60 Å, Supelco®
Supplier:
Merck
About this item
CAS Number:
63231-67-4
:
7631-86-9
Formula:
SiO₂
MDL Number:
MFCD00011232
:
60.08 g/mol
:
M-Clarity™ quality level = MQ400
Storage Temperature:
Ambient