Potassium thiocyanate 0.1 mol/l (0.1 N) in aqueous solution volumetric solution
Catalog # REGEKT201F
Supplier: REAGECON
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
Specifications
About this item
:
:97.18 g/mol
Storage Temperature:Ambient
EINECS:206-370-1
CAS Number:333-20-0
Formula:KSCN
MDL Number:MFCD00011413