Order Entry
Finland
ContactUsLinkComponent
Coomassie® brilliant blue R-250 ~50% (dye content), Technical Grade, Sigma-Aldrich®
Coomassie® brilliant blue R-250 ~50% (dye content), Technical Grade, Sigma-Aldrich®
Certificates
undefined
Coomassie® brilliant blue R-250 ~50% (dye content), Technical Grade, Sigma-Aldrich®

Specification Test Results

  • Appearance (Form):
    Powder
  • Suitability For Detection of Protein Bands Following Electrophoresis:
    Suitable
  • Solubility (Colour):
    Very Dark Purple to Very Dark Blue and Dark Blue-Black
  • Carbon:
    32.0 - 42.0 %
  • Nitrogen:
    2.5 - 3.5 %
  • Dye Content:
    Approximately 50%

Specification Test Results