Cerium(IV) sulphate 0.1 mol/l (0.1 N) in aqueous solution
Catalog # FFCHFF61290200
Supplier: FF CHEMICALS
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:200 mL
- Seq:2530
Specifications
About this item
:
:332.24 g/mol
Merck Index:13,02002
EINECS:237-029-5
UN:3264
ADR:8,III
:GHS07
CAS Number:13590-82-4
Formula:Ce(SO₄)₂
MDL Number:MFCD00148852