Order Entry
Finland
ContactUsLinkComponent
Sudan orange G 85% (dye content), Sigma-Aldrich®
Sudan orange G 85% (dye content), Sigma-Aldrich®
Catalog # 199672-25G
CAS Number:  
undefined
Sudan orange G 85% (dye content), Sigma-Aldrich®
Catalog # 199672-25G
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Powder
  • Infrared spectrum:
    Conforms to Structure
  • Wavelength:
    380 - 420 nm
  • Extinction Coefficient:
    ≥20000
  • Carbon:
    57.2 - 77.4%
  • Nitrogen:
    11.1 - 15.0%
  • Solubility (Turbidity):
    Clear
  • Solubility (Colour):
    Orange to Red

Specification Test Results

About this item

 :
 :214.22 g/mol
Boiling Pt:371 °C (1013 hPa)
Melting Pt:143...146 °C
 :M-Clarity™ quality level = MQ100
Storage Temperature:Room Temperature
EINECS:218-131-9
 :GHS07
CAS Number:2051-85-6
Formula:C₁₂H₁₀N₂O₂
MDL Number:MFCD00002275