Thermo Scientific™ Digital Cel-Gro Tissue Culture Rotator
Catalog # BARN88882016
Supplier: Thermo Fisher Scientific
Specifications
- Temperature:5...40 °F
- Description:Digital Cel-Gro issue culture rotator
- Electrical:100 - 240 V, 50/60Hz, 0,2 A
- Speed range:5 - 70 min⁻¹
- Load capacity:5 kg
- Tilt angle:95 - 180°
- Humidity:20 - 85%
- Speed accuracy:±1 min⁻¹
- Weight:9,5 kg
- W×D×H:260×383×522 mm
- Pk:1
Specifications
About this item
The Thermo Scientific™ Digital Cel-Gro Tissue Culture Rotator precisely mixes test tube contents. Achieve optimum culture results for tissue and virus growth, hormone production, and sample testing of viruses and cytotoxicity assays.
- Digital display of speed and time
- Adjustable rocking angle: 95 - 180°
- Timer range: 0 - 9999 mins
- Optional multiple size drums offering different capacity of tube rotation