Hastelloy C-276 Polarimeter Tube
PAAR109920
New Product
- Capacity:2 ml
- Description:Tube polarimeter, Luer adapter for peltier, 100 mm
- Pk:1
Hastelloy sample cell (polarimeter tube) 100 mm with wireless Toolmaster function which transfers temperature and path length to the MCP 150.
Filling volume approximately 2 ml. Connection to a syringe through Luer filling ports. Suitable for use with Anton Paar polarimeter.