Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,5-Dichloro-2,4,6-trifluorobenzotrifluoride
3,5-Dichloro-2,4,6-trifluorobenzotrifluoride ≥98.0%
3,5-Dichloro-2,4,6-trifluorobenzotrifluoride ≥98.0%
TCIAD2869-5G
:
TCI
:
3,5-Dichloro-2,4,6-trifluorobenzotrifluoride ≥98.0%
TCIAD2869-5G
:
TCI
:
Pk:
5 g
Seq:
3173
Purity/Analysis Method:
>98.0% (GC)
:
:
170 °C
:
4284-10-0
:
C₇Cl₂F₆
:
MFCD00039304