
trans-Cinnamic acid ≥99.8%, zone refined: number of passes 40
Catalog # TCIAC0636-1SAMPLE
Supplier: TCI
CAS Number:
Specifications
- Pk:1
- Seq:0014
Specifications
Specification Test Results
- Purity/Analysis Method:>99.8% (GC)
Specification Test Results
About this item
:
Boiling Pt:300 °C
Melting Pt:135 °C
Density:1.25 g/cm³ (4 °C)
:148.16 g/mol
EINECS:205-398-1
:GHS07
CAS Number:140-10-3
:621-82-9
Formula:C₉H₈O₂
MDL Number:MFCD00004369