EasySep™ HLA Chimerism Whole Blood CD4 Positive Selection Kits
Supplier: STEMCELL TECHNOLOGIES
Certificates
About this item
Column-free immunomagnetic positive selection of human CD4+ cells for HLA analysis.
- Fast and easy-to-use
- Up to 99% purity
- No columns required
The EasySep™ HLA Chimerism Whole Blood CD4 Positive Selection is designed to isolate CD4+ cells from fresh whole blood or buffy coat by positive selection. Desired cells are targeted with Tetrameric Antibody Complexes recognising CD4 and dextran-coated magnetic particles. labelled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off. The CD4 antigen is expressed on helper T cells and more weakly on monocytes and tissue macrophages.
This product replaces the EasySep™ Human Whole Blood CD4 Positive Selection Kit for even faster cell isolations.
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier's mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier's restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').