Order Entry
Finland
ContactUsLinkComponent
EasySep™ Direct Human CTC Enrichment Kit
  STMC19657
undefined
EasySep™ Direct Human CTC Enrichment Kit
  STMC19657
  • Description:
    EasySep™ direct human CTC enrichment kit, 100 ml whole blood processed
  • Pk:
    1 KIT
Immunomagnetic isolation of circulating tumor cells directly from whole blood
  • >99,9% RBC depletion without the need for density gradient centrifugation, sedimentation, or lysis
  • Up to 3 log depletion of normal hematopoietic cells
  • Fast, easy-to-use and column-free
  • Isolated cells are untouched
The EasySep™ Direct Human CTC Enrichment Kit can be used to enrich Circulating Tumor Cells (CTCs) directly from human whole blood by immunomagnetic negative selection without the need for either density gradient centrifugation or RBC lysis. Isolated cells are immediately ready for downstream analysis or assays.
 : Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').