Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4,5-Dibromo-1H-1,2,3-triazole
4,5-Dibromo-1H-1,2,3-triazole 95%
4,5-Dibromo-1H-1,2,3-triazole 95%
:
Thermo Fisher Scientific
New Product
4,5-Dibromo-1H-1,2,3-triazole 95%
:
Thermo Fisher Scientific
New Product
:
15294-81-2
:
:
C₂HBr₂N₃
:
MFCD08062401
:
226.859 g/mol
:
GHS07
:
Refrigerator
Appearance:
white to light yellow powder or crystals
Infrared spectrum:
Conforms
HPLC:
≥94 %