Order Entry
Finland
ContactUsLinkComponent
Methyl orange USP
Methyl orange USP
Catalog # REGEUSP3405
Supplier:  REAGECON
New Product
CAS Number:  
Methyl orange USP
Catalog # REGEUSP3405
Supplier:  REAGECON
New Product
CAS Number:  

Specifications

  • Pack type:
    Amber glass bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

About this item

 :
 :327.34 g/mol
CAS Number:547-58-0
Formula:C₁₄H₁₄N₃NaO₃S
MDL Number:MFCD00007502