System Suitability Set to USP for Total Organic Carbon (TOC) Suitable for Use with Anatel A643 Analyser
Catalog # REGEISTOC1265
Supplier: REAGECON
New Product
CAS Number:
Specifications
- Matrix:H₂O
- Pack type:PP/PE-HD brown bottles
- Pk:1 SET
- Description:Total organic carbon (TOC) standard
- Physical form:Liquid
- Accuracy:±2 %
Specifications
About this item
Reagecon's system suitability set to from USP for Total Organic Carbon (TOC) is a product of extremely high specification and purity. The product is produced from ultra pure raw materials including water that is prepared by a special proprietary process. The product is presented in bottles that are manufactured, cleaned and leached, by a special proprietary process specifically for the product.