0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Finland
ContactUsLinkComponent
N-Octyl-β-D-glucopyranoside
N-Octyl-β-D-glucopyranoside, Millipore®
N-Octyl-β-D-glucopyranoside, Millipore®
:
Merck Millipore (Calbiochem)
N-Octyl-β-D-glucopyranoside, Millipore®
:
Merck Millipore (Calbiochem)
:
29836-26-8
:
:
C₁₄H₂₈O₆
:
MFCD00063288
:
292.37 g/mol
:
110 °C
:
249-887-8
:
GHS07
:
Freezer