Supelco® Organic Reference Standard, Formaldehyde solution
Formaldehyde solution, Stabilised with methanol, ~37 wt.% in H₂O, Certified reference material, 1 mL
Supplier: Merck
Supelco® Organic Reference Standard, Formaldehyde solution
Catalog # 47083-U
Supplier: Merck
CAS Number:
Jump to:
Product Details & Documents
- GHS signal: GHS06, GHS08, GHS05
: P201 - P210 - P280 - P303 + P361 + P353 - P304 + P340 + P310 - P305 + P351 + P338
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Concentration:Stabilised with methanol, ~37 wt.% in H₂O
- Pk:1 mL
- Standards:Certified reference material
- Description:Formaldehyde solution
- Cat. No.:47083-U
- M-Clarity quality level:MQ100
- Formula:HCHO
- Storage conditions:2...30 °C
- Hazard class description:H226 - H301 + H311 - H314 - H317 - H330 - H335 - H341 - H350 - H370
- MDL No.:MFCD00003274
- Flash point:56 °C
- Density:1,09 g/ml at 25 °C (lit.)
- Synonym(s):Formalin
- Molecular weight:30,03
- CAS No.:50-00-0
- Shipping conditions:STD
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



