Order Entry
Finland
ContactUsLinkComponent
Formaldehyde 20% in aqueous solution
Toxic
Health
Formaldehyde 20% in aqueous solution
Catalog # FFCHFF6635000
Supplier:  FF CHEMICALS
CAS Number:  
Toxic
Health
Formaldehyde 20% in aqueous solution
Catalog # FFCHFF6635000
Supplier:  FF CHEMICALS
CAS Number:  

Specifications

  • Pk:
    5 L
  • Seq:
    5190

Specifications

About this item

 :
 :30.03 g/mol
Storage Temperature:Ambient
EINECS:200-001-8
UN:3334
ADR:9,
 :GHS06
 :GHS08
CAS Number:50-00-0
Formula:CH₂O
MDL Number:MFCD00003274