Heptyl β-D-glucopyranoside
Catalog # MOLE66851452-1G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:278.35 g/mol
CAS Number:78617-12-6
Formula:C₁₃H₂₆O₆
MDL Number:MFCD00063299