

Bis(trimethylsilyl)carbodiimide 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1000-70-0
:
Formula:C₇H₁₈N₂Si₂
MDL Number:MFCD00051538
:186.4 g/mol
Boiling Pt:164 °C (1013 hPa)
Melting Pt:–23 °C
Density:0.821 g/cm³
Flash Pt:42 °C
EINECS:213-673-2
UN:1993
:GHS07
:GHS02
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4330 to 1.4350 (20°C, 589 nm)
- GC:≥97.5 %