Order Entry
Finland
Orders LinkContactUsLinkComponent
Cellulase

Cellulase

Glass bottle for solids, 1 g
Catalog # PROOFIP5
Cellulase
Catalog # PROOFIP5
CAS Number:  
Cellulase

Glass bottle for solids, 1 g

CAS Number:  
Price
931,00 €
Log in for availability
1 * 1 g
Product Details & Documents
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Cat. No.:
    PROOFIP5
  • MDL No.:
    MFCD00081510
  • Enzyme name:
    Cellulase
  • CAS No.:
    9012-54-8

Specifications